Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRG15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142185.2 | NP_650442.1 | 3 - 4 | 385 | 66 | |
GenBank mRNA | AF152245.1 | - | 3 - 4 | 370 | 66 | |
AY051679.1 | - | 3 - 4 | 361 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |