Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: h-cup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142205.2 | NP_650462.1 | 1 | 1042 | 123 | |
GenBank mRNA | AY051426.1 | - | 1 | 996 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |