Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pinta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300537.1 | NP_001287466.1 | 1 - 2 | 639 | 73 | |
NM_142785.2 | NP_651042.1 | 1 - 2 | 435 | 73 | ||
GenBank mRNA | BT005203.1 | - | 1 - 2 | 402 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |