Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmdar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169059.2 | NP_730940.1 | 13 - 14 | 2626 | 72 | |
GenBank mRNA | AY070577.1 | - | 13 - 14 | 2622 | 72 | |
X71790.1 | - | 2534 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |