Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTA1-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144539.2 | NP_001138011.1 | 1 | 593 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |