Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144567.2 | NP_001138039.1 | 4 - 5 | 1969 | 61 | |
NM_001300368.1 | NP_001287297.1 | 4 - 5 | 2785 | 61 | ||
NM_001300369.1 | NP_001287298.1 | 4 - 5 | 1969 | 61 | ||
NM_169451.2 | NP_731670.1 | 5 - 6 | 2791 | 61 | ||
NM_169452.3 | NP_731671.1 | 4 - 5 | 1969 | 61 | ||
GenBank mRNA | BT133437.1 | - | 5 - 6 | 2789 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |