Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LpR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104461.3 | NP_001097931.3 | 9 - 10 | 2719 | 64 | |
NM_001104462.1 | NP_001097932.1 | 8 - 9 | 2548 | 64 | ||
NM_001104463.3 | NP_001097933.2 | 9 - 10 | 3031 | 64 | ||
NM_001104464.2 | NP_001097934.2 | 11 - 12 | 3271 | 64 | ||
NM_001170263.2 | NP_001163734.1 | 9 - 10 | 2977 | 64 | ||
NM_001276043.1 | NP_001262972.1 | 9 - 10 | 2971 | 64 | ||
NM_001276044.1 | NP_001262973.1 | 10 - 11 | 5191 | 64 | ||
NM_001276045.1 | NP_001262974.1 | 8 - 9 | 2465 | 64 | ||
NM_170240.3 | NP_733119.2 | 9 - 10 | 2725 | 64 | ||
NM_206573.2 | NP_996296.2 | 10 - 11 | 3154 | 64 | ||
GenBank mRNA | BT011057.1 | - | 9 - 10 | 3031 | 64 | |
BT011063.1 | - | 10 - 11 | 3133 | 64 | ||
BT099946.1 | - | 8 - 9 | 2548 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |