Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202364.1 | NP_001189293.1 | 4 - 5 | 720 | 74 | |
NM_001202365.2 | NP_001189294.1 | 4 - 5 | 714 | 74 | ||
NM_001260380.2 | NP_001247309.1 | 4 - 5 | 714 | 74 | ||
NM_001276052.1 | NP_001262981.1 | 4 - 5 | 714 | 74 | ||
NM_001300610.1 | NP_001287539.1 | 4 - 5 | 714 | 74 | ||
NM_079790.5 | NP_524514.2 | 4 - 5 | 653 | 74 | ||
NM_170254.3 | NP_733133.1 | 4 - 5 | 733 | 74 | ||
NM_170255.4 | NP_733134.1 | 4 - 5 | 714 | 74 | ||
NM_170256.2 | NP_733135.1 | 4 - 5 | 632 | 74 | ||
NM_206575.3 | NP_996298.1 | 4 - 5 | 714 | 74 | ||
GenBank mRNA | AF145695.1 | - | 4 - 5 | 699 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |