Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mfas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176478.1 | NP_788655.1 | 3 - 4 | 256 | 66 | |
GenBank mRNA | BT133296.1 | - | 3 - 4 | 235 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |