Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300403.1 | NP_001287332.1 | 3 - 4 | 1130 | 95 | |
NM_206493.2 | NP_996215.1 | 3 - 4 | 1130 | 95 | ||
GenBank mRNA | AY102651.1 | - | 3 - 4 | 1112 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |