Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274942.1 | NP_001261871.1 | 2 | 1441 | 116 | |
NM_140496.3 | NP_648753.1 | 2 | 1427 | 116 | ||
GenBank mRNA | AY051934.1 | - | 2 | 1414 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |