Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kermit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299258.1 | NP_001286187.1 | 1 | 481 | 62 | |
NM_143771.4 | NP_652028.1 | 2 | 898 | 62 | ||
NM_165590.3 | NP_724649.1 | 2 | 539 | 62 | ||
NM_165591.3 | NP_724650.1 | 2 | 542 | 62 | ||
NM_165592.3 | NP_724651.1 | 2 | 426 | 62 | ||
GenBank mRNA | AY060451.1 | - | 2 | 819 | 62 | |
AY095011.1 | - | 2 | 373 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |