Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014589.4 | NP_001014589.1 | 3 - 4 | 651 | 67 | |
NM_001038939.2 | NP_001034028.2 | 3 - 4 | 651 | 67 | ||
GenBank mRNA | BT031129.1 | - | 3 - 4 | 617 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |