Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nplp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272964.2 | NP_001259893.1 | 2 | 272 | 64 | |
NM_144095.3 | NP_652352.1 | 2 | 230 | 64 | ||
GenBank mRNA | BT088864.1 | - | 2 | 225 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |