Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipped-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042986.3 | NP_001036451.1 | 12 - 13 | 3683 | 122 | |
NM_001042987.3 | NP_001036452.1 | 12 - 13 | 3711 | 122 | ||
NM_001169581.2 | NP_001163052.1 | 13 - 14 | 3604 | 122 | ||
NM_001169582.2 | NP_001163053.1 | 13 - 14 | 3639 | 122 | ||
NM_001169583.2 | NP_001163054.1 | 13 - 14 | 3611 | 122 | ||
NM_001273774.1 | NP_001260703.1 | 11 - 12 | 3351 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |