Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stlk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042977.3 | NP_001036442.1 | 1 - 2 | 362 | 129 | |
NM_001259209.2 | NP_001246138.1 | 1 - 2 | 362 | 129 | ||
GenBank mRNA | AY069348.1 | - | 1 - 2 | 333 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |