Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002162.1 | NP_001002162.1 | 2 - 3 | 306 | 140 | |
GenBank mRNA | BC071305.1 | - | 2 - 3 | 306 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |