Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002523.2 | NP_001002523.1 | 3 - 4 | 515 | 87 | |
XM_005156815.4 | XP_005156872.1 | 4 - 5 | 579 | 87 | ||
XM_005156816.4 | XP_005156873.1 | 3 - 4 | 542 | 87 | ||
GenBank mRNA | BC076274.1 | - | 3 - 4 | 516 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |