Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eef2kmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006058.1 | NP_001006058.1 | 7 - 8 | 948 | 100 | |
XM_009299451.2 | XP_009297726.1 | 7 - 8 | 1074 | 100 | ||
GenBank mRNA | BC083266.1 | - | 7 - 8 | 948 | 100 | |
BC090275.1 | - | 2 - 3 | 1196 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |