Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phactr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020537.1 | NP_001018373.1 | 5 - 6 | 794 | 73 | |
XM_021479617.1 | XP_021335292.1 | 5 - 6 | 804 | 73 | ||
GenBank mRNA | BC095090.1 | - | 5 - 6 | 794 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |