Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: siae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044331.1 | NP_001037796.1 | 6 - 7 | 843 | 81 | |
GenBank mRNA | BC049448.1 | - | 6 - 7 | 843 | 81 | |
BC134862.1 | - | 6 - 7 | 815 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |