Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: axdnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089377.1 | NP_001082846.1 | 20 - 21 | 2628 | 90 | |
GenBank mRNA | BC139551.1 | - | 20 - 21 | 2628 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |