Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130933.2 | NP_571008.1 | 2 - 3 | 1402 | 62 | |
GenBank mRNA | AF052435.1 | - | 2 - 3 | 62 | ||
AF127981.1 | - | 2 - 3 | 62 | |||
BC066379.1 | - | 2 - 3 | 62 | |||
BC115246.1 | - | 2 - 3 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |