Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam49ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200070.3 | NP_956364.2 | 9 - 10 | 1235 | 69 | |
GenBank mRNA | AY394961.1 | - | 9 - 10 | 935 | 69 | |
DT055468.1 | - | 2 - 3 | 547 | 69 | ||
EH507686.1 | - | 5 - 6 | 636 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |