Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbgt1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200329.1 | NP_956623.1 | 4 - 5 | 259 | 151 | |
GenBank mRNA | BC052142.1 | - | 4 - 5 | 259 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |