Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201454.2 | NP_958862.1 | 2 - 3 | 347 | 139 | |
GenBank mRNA | BC044459.1 | - | 2 - 3 | 202 | 139 | |
BC165141.1 | - | 2 - 3 | 151 | 139 | ||
EH456193.1 | - | 2 - 3 | 319 | 139 | ||
EH467102.1 | - | 6 - 7 | 678 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |