Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyb5d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017684.2 | NP_001017684.1 | 1 - 2 | 201 | 80 | |
GenBank mRNA | BC093157.1 | - | 1 - 2 | 204 | 80 | |
BC155725.1 | - | 1 - 2 | 213 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |