Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loxl5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045856.2 | NP_001039321.2 | 3 - 4 | 1091 | 140 | |
XM_021469354.1 | XP_021325029.1 | 3 - 4 | 1195 | 140 | ||
GenBank mRNA | EF030485.1 | - | 3 - 4 | 1091 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |