Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plcd3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099423.1 | NP_001092893.1 | 14 - 15 | 2610 | 72 | |
GenBank mRNA | BC139849.1 | - | 14 - 15 | 2610 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |