Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lhx1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131216.1 | NP_571291.1 | 2 - 3 | 424 | 77 | |
GenBank mRNA | BC162396.1 | - | 2 - 3 | 420 | 77 | |
BC162664.1 | - | 2 - 3 | 420 | 77 | ||
L37802.1 | - | 2 - 3 | 424 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |