Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201099.1 | NP_957393.1 | 3 - 4 | 459 | 76 | |
XM_021476043.1 | XP_021331718.1 | 3 - 4 | 76 | |||
GenBank mRNA | BC045336.1 | - | 3 - 4 | 459 | 76 | |
BC164693.1 | - | 3 - 4 | 471 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |