Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002086.1 | NP_001002086.1 | 6 - 7 | 519 | 153 | |
GenBank mRNA | BC071544.1 | - | 6 - 7 | 519 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |