Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pacsin1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328357.1 | NP_001315286.1 | 2 - 3 | 276 | 59 | |
XM_005161905.4 | XP_005161962.1 | 2 - 3 | 256 | 59 | ||
XM_021469807.1 | XP_021325482.1 | 2 - 3 | 257 | 59 | ||
GenBank mRNA | BC095091.1 | - | 2 - 3 | 244 | 59 | |
BC164592.1 | - | 1 - 2 | 91 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |