Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smtla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037706.1 | NP_001032795.1 | 2 - 3 | 258 | 68 | |
XM_021467699.1 | XP_021323374.1 | 2 - 3 | 249 | 68 | ||
GenBank mRNA | AY376857.1 | - | 2 - 3 | 258 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |