Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slmapa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077254.1 | NP_001070722.1 | 19 - 20 | 2572 | 83 | |
XM_005168060.4 | XP_005168117.1 | 20 - 21 | 3328 | 83 | ||
XM_005168061.4 | XP_005168118.1 | 20 - 21 | 3328 | 83 | ||
XM_005168062.4 | XP_005168119.1 | 19 - 20 | 3265 | 83 | ||
XM_005168063.4 | XP_005168120.1 | 18 - 19 | 3214 | 83 | ||
XM_005168064.4 | XP_005168121.1 | 18 - 19 | 1934 | 83 | ||
GenBank mRNA | BC115271.1 | - | 9 - 10 | 924 | 83 | |
BC124763.1 | - | 19 - 20 | 2572 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |