Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp3c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077548.1 | NP_001071016.1 | 3 - 4 | 286 | 146 | |
GenBank mRNA | BC124295.1 | - | 3 - 4 | 286 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |