Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: entpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083581.1 | NP_001077050.1 | 4 - 5 | 445 | 89 | |
XM_005157870.2 | XP_005157927.1 | 4 - 5 | 532 | 89 | ||
GenBank mRNA | BC134912.1 | - | 4 - 5 | 445 | 89 | |
EF446129.1 | - | 4 - 5 | 450 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |