Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089361.1 | NP_001082830.1 | 7 - 8 | 785 | 69 | |
XM_005172508.4 | XP_005172565.1 | 7 - 8 | 769 | 69 | ||
GenBank mRNA | BC135034.1 | - | 7 - 8 | 785 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |