Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbxta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131162.1 | NP_571237.1 | 6 - 7 | 963 | 90 | |
GenBank mRNA | BC162297.1 | - | 6 - 7 | 958 | 90 | |
BC162300.1 | - | 6 - 7 | 958 | 90 | ||
S57147.1 | - | 6 - 7 | 963 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |