Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tlcd3ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199637.1 | NP_955931.1 | 5 - 6 | 669 | 85 | |
XM_005163853.4 | XP_005163910.1 | 6 - 7 | 871 | 85 | ||
XM_005163854.4 | XP_005163911.1 | 6 - 7 | 860 | 85 | ||
XM_005163855.4 | XP_005163912.1 | 7 - 8 | 927 | 85 | ||
XM_005163856.4 | XP_005163913.1 | 7 - 8 | 924 | 85 | ||
XM_005163857.4 | XP_005163914.1 | 6 - 7 | 867 | 85 | ||
XM_005163858.4 | XP_005163915.1 | 6 - 7 | 855 | 85 | ||
XM_005163859.4 | XP_005163916.1 | 6 - 7 | 856 | 85 | ||
XM_005163860.4 | XP_005163917.1 | 5 - 6 | 1057 | 85 | ||
XM_017353338.2 | XP_017208827.1 | 7 - 8 | 884 | 85 | ||
XM_021469449.1 | XP_021325124.1 | 7 - 8 | 960 | 85 | ||
XM_021469458.1 | XP_021325133.1 | 5 - 6 | 667 | 85 | ||
GenBank mRNA | BC059567.1 | - | 5 - 6 | 669 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |