Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003436.2 | NP_001003436.1 | 2 - 3 | 268 | 129 | |
GenBank mRNA | BC076312.1 | - | 2 - 3 | 117 | 129 | |
BC164444.1 | - | 2 - 3 | 140 | 129 | ||
EH500639.1 | - | 2 - 3 | 159 | 129 | ||
EH548721.1 | - | 2 - 3 | 308 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |