Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nup85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003625.1 | NP_001003625.1 | 717 | 63 | ||
GenBank mRNA | BC078318.1 | - | 717 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio |