Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myo6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004111.2 | NP_001004111.2 | 29 - 30 | 3185 | 74 | |
XM_005160309.4 | XP_005160366.1 | 30 - 31 | 3331 | 74 | ||
XM_009294500.3 | XP_009292775.1 | 31 - 32 | 3340 | 74 | ||
XM_021468402.1 | XP_021324077.1 | 31 - 32 | 3340 | 74 | ||
XM_021468403.1 | XP_021324078.1 | 29 - 30 | 3179 | 74 | ||
XM_021468404.1 | XP_021324079.1 | 30 - 31 | 3331 | 74 | ||
GenBank mRNA | AY618702.1 | - | 29 - 30 | 3183 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |