Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ankrd45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020770.1 | NP_001018606.1 | 3 - 4 | 545 | 115 | |
GenBank mRNA | BC095811.1 | - | 3 - 4 | 545 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |