Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sycp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040350.1 | NP_001035440.1 | 7 - 8 | 824 | 105 | |
GenBank mRNA | BC115343.1 | - | 7 - 8 | 824 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |