Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clcn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082910.1 | NP_001076379.1 | 6 - 7 | 683 | 133 | |
XM_009294060.3 | XP_009292335.1 | 6 - 7 | 1014 | 133 | ||
XM_017352282.2 | XP_017207771.1 | 5 - 6 | 885 | 133 | ||
XM_021468144.1 | XP_021323819.1 | 6 - 7 | 786 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |