Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: puf60a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105588.1 | NP_001099058.1 | 10 - 11 | 1162 | 84 | |
XM_005163500.4 | XP_005163557.1 | 10 - 11 | 1169 | 84 | ||
XM_005163501.4 | XP_005163558.1 | 10 - 11 | 1156 | 84 | ||
XM_005163502.4 | XP_005163559.1 | 11 - 12 | 1226 | 84 | ||
XM_005163503.3 | XP_005163560.1 | 11 - 12 | 1296 | 84 | ||
XM_005163505.3 | XP_005163562.1 | 12 - 13 | 1334 | 84 | ||
XM_005163506.3 | XP_005163563.1 | 12 - 13 | 1387 | 84 | ||
XM_005163507.4 | XP_005163564.1 | 12 - 13 | 1386 | 84 | ||
XM_005163508.4 | XP_005163565.1 | 11 - 12 | 1275 | 84 | ||
XM_005163509.4 | XP_005163566.1 | 9 - 10 | 1104 | 84 | ||
XM_005163510.1 | XP_005163567.1 | 7 - 8 | 824 | 84 | ||
XM_009298620.3 | XP_009296895.1 | 12 - 13 | 1333 | 84 | ||
XM_009298622.3 | XP_009296897.1 | 11 - 12 | 1276 | 84 | ||
XM_017358672.2 | XP_017214161.1 | 5 - 6 | 496 | 84 | ||
GenBank mRNA | BC135083.1 | - | 10 - 11 | 1174 | 84 | |
BC153301.1 | - | 10 - 11 | 1162 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |