Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shisa9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013509.1 | NP_001013527.1 | 2 - 3 | 895 | 82 | |
GenBank mRNA | BC090494.1 | - | 2 - 3 | 895 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |