Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131292.1 | NP_571367.1 | 14 - 15 | 978 | 83 | |
XM_005166863.4 | XP_005166920.1 | 13 - 14 | 910 | 83 | ||
GenBank mRNA | BC163934.1 | - | 14 - 15 | 970 | 83 | |
BC167477.1 | - | 13 - 14 | 874 | 83 | ||
BC168504.1 | - | 14 - 15 | 1059 | 83 | ||
DQ335127.1 | - | 14 - 15 | 978 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |