Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: laptm4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200098.2 | NP_956392.1 | 3 - 4 | 470 | 100 | |
GenBank mRNA | BC059484.1 | - | 3 - 4 | 399 | 100 | |
EH440716.1 | - | 3 - 4 | 443 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |